Merck
  • Important role of Maillard reaction in the protective effect of heat-processed ginsenoside Re-serine mixture against cisplatin-induced nephrotoxicity in LLC-PK1 cells.
Important role of Maillard reaction in the protective effect of heat-processed ginsenoside Re-serine mixture against cisplatin-induced nephrotoxicity in LLC-PK1 cells.
Bioorganic & medicinal chemistry letters (2012-08-02)
Ji Hwan Lee, Woojung Lee, Seungyong Lee, Yujung Jung, Soon-Hye Park, Pilju Choi, Su-Nam Kim, Jungyeob Ham, Ki Sung Kang
ABSTRACT
The aim of the present study was to verify the important role of Maillard reaction in the protective effect of heat-processed ginsenoside Re-serine mixture against oxidative stress-induced nephrotoxicity. The free radical-scavenging activity of ginsenoside Re-serine mixture was increased by heat-processing. Ginsenoside Re was transformed into less-polar ginsenosides such as Rg(2), Rg(6) and F(4) by heat-processing, and the glucose molecule at carbon-20 was separated. The improved-free radical-scavenging activity by heat-processing was mediated by the generation of antioxidant Maillard reaction products (MRPs) from the reaction of glucose with serine. Moreover, MRPs from ginsenoside Re-serine mixture showed protective effect against cisplatin-induced renal epithelial cell damage.
Product Description
Ginsenoside Re, primary reference standard