Merck
Short synthesis of diamide-linked sucrose macrocycles.
Organic letters (2012-08-03)
Mykhaylo A Potopnyk, Piotr Cmoch, Sławomir Jarosz
ABSTRACT
A convenient route to macrocyclic diamide-linked macrocyclic derivatives with a sucrose scaffold is presented. Reaction of sucrose based amines (o- and m-) with acid dichlorides afforded the monomeric macrocycles in excellent yields, while reaction of the p-amines also provided dimeric products.
Sigma-Aldrich
Diamide