Merck
  • Rapid and selective quantification of L-theanine in ready-to-drink teas from Chinese market using SPE and UPLC-UV.
Rapid and selective quantification of L-theanine in ready-to-drink teas from Chinese market using SPE and UPLC-UV.
Food chemistry (2012-08-08)
Guoqiang Chen, Yun Wang, Weiqi Song, Bo Zhao, Yuling Dou
ABSTRACT
An ultra performance liquid chromatography (UPLC) method combined with solid phase extraction (SPE) sample pre-treatment was developed and validated for the rapid quantification of L-theanine in ready-to-drink (RTD) teas. UPLC analysis of twenty-seven RTD teas from the Chinese market revealed that the L-theanine levels in various types of RTD teas were significantly different. RTD green teas were found to contain highest mean L-theanine level (37.85±20.54 mg/L), followed by jasmine teas (36.60±12.08 mg/L), Tieguanying teas (18.54±3.46 mg/L) black teas (16.89±6.56), Pu-erh teas (11.31±0.90 mg/L) and oolong teas (3.85±2.27 mg/L). The ratio of total polyphenols content to L-theanine content could be used as a featured parameter for differentiating RTD teas. L-theanine in RTD teas could be a reliable quality parameter that is complementary to total polyphenols.