Merck
  • Assessment of effective absorbed dose of (111)In-DTPA-Buserelin in human on the basis of biodistribution rat data.
Assessment of effective absorbed dose of (111)In-DTPA-Buserelin in human on the basis of biodistribution rat data.
Radiation protection dosimetry (2012-08-10)
Afsaneh Lahooti, Saeed Shanehsazzadeh, Amir Reza Jalilian, Mohammad Bagher Tavakoli
ABSTRACT
In this study, the effective absorbed dose to human organs was estimated, following intra vascular administration of (111)In-DTPA-Buserelin using biodistribution data from rats. Rats were sacrificed at exact time intervals of 0.25, 0.5, 1, 2, 4 and 24 h post injections. The Medical Internal Radiation Dose formulation was applied to extrapolate from rats to humans and to project the absorbed radiation dose for various human organs. From rat data, it was estimated that a 185-MBq injection of (111)In-DTPA-Buserelin into the human might result in an estimated absorbed dose of 24.27 mGy to the total body and the highest effective absorbed dose was in kidneys, 28.39 mSv. The promising results of this study emphasises the importance of absorbed doses in humans estimated from data on rats.
Sigma-Aldrich
Buserelin acetate salt, ≥90% (HPLC)