Merck
  • Skin phototoxicity of cosmetic formulations containing photounstable and photostable UV-filters and vitamin A palmitate.
Skin phototoxicity of cosmetic formulations containing photounstable and photostable UV-filters and vitamin A palmitate.
Toxicology in vitro : an international journal published in association with BIBRA (2012-08-22)
Lorena R Gaspar, Julian Tharmann, Patricia M B G Maia Campos, Manfred Liebsch
ABSTRACT
The aim of this study was to evaluate the in vitro skin phototoxicity of cosmetic formulations containing photounstable and photostable UV-filters and vitamin A palmitate, assessed by two in vitro techniques: 3T3 Neutral Red Uptake Phototoxicity Test and Human 3-D Skin Model In Vitro Phototoxicity Test. For this, four different formulations containing vitamin A palmitate and different UV-filters combinations, two of them considered photostable and two of them considered photounstable, were prepared. Solutions of each UV-filter and vitamin under study and solutions of four different combinations under study were also prepared. The phototoxicity was assessed in vitro by the 3T3 NRU phototoxicity test (3T3-NRU-PT) and subsequently in a phototoxicity test on reconstructed human skin model (H3D-PT). Avobenzone presented a pronounced phototoxicity and vitamin A presented a tendency to a weak phototoxic potential. A synergistic effect of vitamin A palmitate on the phototoxicity of combinations containing avobenzone was observed. H3D-PT results did not confirm the positive 3T3-NRU-PT results. However, despite the four formulations studied did not present any acute phototoxicity potential, the combination 2 containing octyl methoxycinnamate (OMC), avobenzone (AVB) and 4-methylbenzilidene camphor (MBC) presented an indication of phototoxicity that should be better investigated in terms of the frequency of photoallergic or chronic phototoxicity in humans, once these tests are scientifically validated only to detect phototoxic potential with the aim of preventing phototoxic reactions in the general population, and positive results cannot predict the exact incidence of phototoxic reactions in humans.
Sigma-Aldrich
Retinyl palmitate, potency: ≥1,700,000 USP units per g
Sigma-Aldrich
Retinyl palmitate, Type IV, ~1,800,000 USP units/g, oil
Sigma-Aldrich
Neutral Red Solution (0.33%), 3.3 g/L in DPBS, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Neutral Red powder, BioReagent, cell culture | mammalian | 553-24-2, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Neutral Red, Dye content ≥90 %
Sigma-Aldrich
Neutral Red, suitable for microscopy
Sigma-Aldrich
Neutral Red, certified by the BSC
Supelco
Retinol palmitate, analytical standard
Sigma-Aldrich
Neutral Red Solution, Buffered
Supelco
Retinyl Palmitate (Vitamin A Palmitate), Pharmaceutical Secondary Standard; Certified Reference Material