Merck
  • Exposure-response relationships in asthma caused by tetrachlorophthalic anhydride.
Exposure-response relationships in asthma caused by tetrachlorophthalic anhydride.
The Journal of allergy and clinical immunology (1990-01-01)
K M Venables, A J Newman Taylor
ABSTRACT
Four patients with asthma caused by occupational exposure to tetrachlorophthalic anhydride (TCPA) had dust-challenge tests that used eight different levels of TCPA exposure. Measurements of TCPA in the test-cubicle air ranged from 1.3 to 961.1 micrograms/m3. The higher exposures provoked definite late or dual asthmatic responses. Smaller, but statistically significant, responses were observed at low exposures. The late asthmatic-response area had a linear relation to the logarithm of TCPA exposure, but the immediate asthmatic response was less closely related to exposure.
Sigma-Aldrich
Tetrachlorophthalic anhydride, 96%