Merck
  • Assessing oxazole bioisosteres as mutasynthons on the rhizoxin assembly line.
Assessing oxazole bioisosteres as mutasynthons on the rhizoxin assembly line.
Chembiochem : a European journal of chemical biology (2012-10-30)
Björn Kusebauch, Nicole Brendel, Heike Kirchner, Hans-Martin Dahse, Christian Hertweck
ABSTRACT
Chain armor against tumor cells: The oxazole side chain in the antimitotic agent rhizoxin S2 (1) was successfully replaced through mutasynthesis by using an engineered mutant impaired in heterocyclization. Incorporation of 12 non-natural surrogates into fully processed rhizoxin analogues revealed a remarkable substrate flexibility of the PKS-NRPS hybrid.