Merck
  • Synthesis and evaluation of a series of 3,4,5-trimethoxycinnamic acid derivatives as potential antinarcotic agents.
Synthesis and evaluation of a series of 3,4,5-trimethoxycinnamic acid derivatives as potential antinarcotic agents.
Chemical biology & drug design (2012-11-06)
Jae-Chul Jung, Sohyeon Moon, Dongguk Min, Woo Kyu Park, Mankil Jung, Seikwan Oh
ABSTRACT
A series of 3,4,5-trimethoxycinnamic acid derivatives was prepared and evaluated for antinarcotic effects on morphine dependence in mice and binding affinities on serotonergic receptors. The key synthetic strategies involve generation of ketones 6-7, esters 9-12 through condensation reaction, and amides 13-19 via coupling reaction using 1-hydroxybenzotriazole/ethyl(dimethylaminopropryl)carbodiimide system in high yield. We found that the naloxone-induced morphine withdrawal syndrome was significantly suppressed by new synthetic 3,4,5-trimethoxycinnamic acid derivatives (20 mg/kg/day). Most of 3,4,5-trimethoxycinnamic acid derivatives were found to have high affinity to 5-HT(1A) receptor. The naloxone-induced morphine withdrawal syndrome was attenuated by (+)8-OH-DPAT (0.1 mg/kg/day, i.p.), a 5-HT(1A) receptor agonist. In cortical neuronal cells, (+)8-OH-DPAT (1 μM) produced an elevation of the pERK 1/2 expression, and the elevated pERK levels were inhibited by WAY 100635, a 5-HT(1A) receptor-specific antagonist. Interestingly, the pERK levels were increased by the 3,4,5-trimethoxycinnamic acid derivatives and the derivatives-mediated changes in pERK levels were blocked by the WAY 100635. These results suggested that new synthetic 3,4,5-trimethoxycinnamic acid derivatives have a potential antinarcotic effect through acting as a 5-HT(1A) receptor agonist in mice.