Merck
  • Liquid chromatographic determination of microcystins in water samples following pre-column excimer fluorescence derivatization with 4-(1-pyrene)butanoic acid hydrazide.
Liquid chromatographic determination of microcystins in water samples following pre-column excimer fluorescence derivatization with 4-(1-pyrene)butanoic acid hydrazide.
Analytica chimica acta (2012-11-14)
Tadashi Hayama, Kenji Katoh, Takayoshi Aoki, Miki Itoyama, Kenichiro Todoroki, Hideyuki Yoshida, Masatoshi Yamaguchi, Hitoshi Nohta
ABSTRACT
A method to measure the concentrations of microcystins (MCs) in water samples has been developed by incorporating pre-column fluorescence derivatization and liquid chromatography (LC). A solid-phase extraction for pretreatment was used to extract the MCs in water samples. The MCs were derivatized with excimer-forming 4-(1-pyrene)butanoic acid hydrazide (PBH). The MCs could then be detected by fluorescence after separation with a pentafluorophenyl (PFP)-modified superficially porous (core shell) particle LC column. The derivatization reactions of MCs with PBH proceeded easily in the presence of 4,6-dimethoxy-1,3,5-triazin-2-yl-4-methylmorpholinium (DMT-MM) as a condensation reagent, and the resulting derivatives could be easily separated on the PFP column. The derivatives were selectively detected at excimer fluorescence wavelengths (440-540 nm). The instrument detection limit and the instrument quantification limit of the MCs standards were 0.4-1.2 μg L(-1) and 1.4-3.9 μg L(-1), respectively. The method was validated at 0.1 and 1.0 μg L(-1) levels in tap and pond water samples, and the recovery of MCs was between 67 and 101% with a relative standard deviation of 11%. The proposed method can be used to quantify trace amounts of MCs in water samples.
Sigma-Aldrich
1-Pyrenebutyric hydrazide, suitable for fluorescence, ≥97.0% (T)