Merck
  • [Effect of di-(2-ethylhexyl)phthalate and its metabolite mono(2-ethylhexyl)phthalate on spermatogenic cell apoptosis in young male Wistar rats].
[Effect of di-(2-ethylhexyl)phthalate and its metabolite mono(2-ethylhexyl)phthalate on spermatogenic cell apoptosis in young male Wistar rats].
Nan fang yi ke da xue xue bao = Journal of Southern Medical University (2012-12-27)
Junjie Yang, Hong Ma, Jing Li, Hong Liu, Weitong Zhang, Yongzheng Zhou, Peng Zhao
ABSTRACT
To explore the influences of di-(2-ethylhexyl)phthalate (DEHP) and its principle metabolite mono(2-ethylhexyl)phthalate (MEHP) on spermatogenic cell apoptosis in young male Wistar rats. Ninety-eight 2-week-old male Wistar rats were randomly divided into 14 equal groups to receive daily intragastric administration of 0.2 ml/kg normal saline for 3 weeks (normal control), 100 mg/kg cyclophosphamide (CTX) for 1 week (positive control), 100, 200, and 300 mg/kg DEHP or MEHP for 1 week, or 100 mg/kg DEHP or MEHP for 1, 2, and 3 weeks. After the treatments, the pathological changes of the testicular tissues were examined, spermatogenic cell apoptosis was detected, and serum sex hormones levels were measured using TUNEL assay or radioimmunoassays. CTX, DEHP, and MEHP all caused shrinkage, development retardation and quantitative reduction of spermatogenic cells with and mitochondrial swelling vacuolar changes. The damage of spermatogenic cells increased significantly with the increment of DEHP and MEHP doses and exposure time. Both DEHP and MEHP treatments resulted in significantly increased cell apoptosis index (AI) in close correlation with the exposure doses and duration (P<0.01). DEHP and MEHP treatments also significantly increased serum levels of follicle stimulating hormone and luteinizing hormone and decreased testosterone levels in a dose- and time-dependent manner (P<0.05). DEHP and MEHP can induce obvious apoptosis of spermatogenic cells in young male rats with a dose- and time-dependent effect.
Product Description
Supelco
Bis(2-ethylhexyl) phthalate solution, certified reference material, 2000 μg/mL in methanol
Supelco
Bis(2-ethylhexyl) phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Bis(2-ethylhexyl) phthalate, Selectophore
Sigma-Aldrich
Dioctyl phthalate, ≥99.5%
Supelco
Bis(2-ethylhexyl) phthalate, PESTANAL®, analytical standard