Merck
  • Sterol regulatory element binding transcription factor 1 (SREBF1) polymorphism and milk fatty acid composition.
Sterol regulatory element binding transcription factor 1 (SREBF1) polymorphism and milk fatty acid composition.
Journal of dairy science (2013-02-14)
R A Nafikov, J P Schoonmaker, K T Korn, K Noack, D J Garrick, K J Koehler, J Minick-Bormann, J M Reecy, D E Spurlock, D C Beitz
ABSTRACT
Milk is known to contain high concentrations of saturated fatty acids-such as palmitic (16:0), myristic (14:0), and lauric (12:0) acids-that can raise plasma cholesterol in humans, making their presence in milk undesirable. The main objective of our candidate gene study was to develop genetic markers that can be used to improve the healthfulness of bovine milk. The sterol regulatory element binding transcription factor 1 (SREBF1) known to regulate the transcription of lipogenic genes together with SREBF chaperone and insulin induced gene 1 were the candidate genes. The results showed significant association of the overall SREBF1 haplotypes with milk production and variations in lauric (12:0) and myristic (14:0) acid concentrations in milk. Haplotype H1 of SREBF1 was the most desirable to improve milk healthfulness because it was significantly associated with lower lauric (12:0) and myristic (14:0) acid concentrations compared with haplotype H3 of SREBF1, and lower lauric acid (12:0) concentration compared with haplotype H2 of SREBF1. Haplotype H1 of SREBF1, however, was significantly associated with lower milk production compared with haplotype H3 of SREBF1. We did not detect any significant associations between genetic polymorphisms in insulin induced gene 1 (INSIG1) and SREBF chaperone and milk fatty acid composition. In conclusion, genetic polymorphisms in SREBF1 can be used to develop genetic tools for the selection of animals producing milk with healthier fatty acid composition.
Product Description
Supelco
Myristic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium dodecanoate, 99-100%
Sigma-Aldrich
Lauric acid, natural, ≥98%, FCC, FG
Sigma-Aldrich
Lauric acid, ≥98%, FCC, FG
Sigma-Aldrich
Myristic acid, natural, ≥98.5%, FG
Sigma-Aldrich
Myristic acid, ≥95%, FCC, FG
Supelco
Myristic acid, analytical standard
Sigma-Aldrich
Myristic acid, ≥98.0% (GC)
Sigma-Aldrich
Myristic acid, Sigma Grade, ≥99%
Supelco
Dodecanoic acid, analytical standard
Sigma-Aldrich
Dodecanoic acid, 98%
Sigma-Aldrich
Dodecanoic acid, ≥99% (GC/titration)