Merck
  • Photoelectrochemical properties of highly mobilized Li-doped ZnO thin films.
Photoelectrochemical properties of highly mobilized Li-doped ZnO thin films.
Journal of photochemistry and photobiology. B, Biology (2013-02-19)
S S Shinde, C H Bhosale, K Y Rajpure
ABSTRACT
Li-doped ZnO thin films with preferred (002) orientation have been prepared by spray pyrolysis technique in aqueous medium on to the corning glass substrates. The effect of Li-doping on to the photoelectrochemical, structural, morphological, optical, luminescence, electrical and thermal properties has been investigated. XRD and Raman study indicates that the films have hexagonal crystal structure. The transmittance, reflectance, refractive index, extinction coefficient and bandgap have been analyzed by optical study. PL spectra consist of a near band edge and visible emission due to the electronic defects, which are related to deep level emissions, such as oxide antisite (OZn), interstitial zinc (Zni), interstitial oxygen (Oi) and zinc vacancy (VZn). The Li-doped ZnO films prepared for 1at% doping possesses the highest electron mobility of 102cm(2)/Vs and carrier concentration of 3.62×10(19)cm(-3). Finally, degradation of 2,4,6-Trinitrotoluene using Li-doped ZnO thin films has been reported.
Product Description
Supelco
2,4,6-Trinitrotoluene solution, 1000 μg/mL in acetonitrile, ampule of 1.2 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.
Supelco
2,4,6-Trinitrotoluene solution, 10 mg/mL in acetonitrile, ampule of 5 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.