Merck
  • One-pot syntheses of isoquinolin-3-ones and benzo-1,4-diazepin-2,5-diones utilizing Ugi-4CR post-transformation strategy.
One-pot syntheses of isoquinolin-3-ones and benzo-1,4-diazepin-2,5-diones utilizing Ugi-4CR post-transformation strategy.
ACS combinatorial science (2013-02-28)
Chao Che, Song Li, Zhixiong Yu, Fangfang Li, Shengchang Xin, Liyan Zhou, Shuo Lin, Zhen Yang
ABSTRACT
One-pot and efficient syntheses of structurally diverse isoquinolin-3-ones and isoquinolin-3-one-based benzo-1,4-diazepin-2,5-diones have been developed. The notable features of the process include the Ugi condensation of monomasked phthalaldehydes with amines, carboxylic acids, and isonitriles, followed by HClO4-mediated intramolecular condensation of the carbonyl with amide.
Sigma-Aldrich
Phthaldialdehyde, suitable for HPLC fluorimetric detection of amino acids, ≥99% (HPLC), powder or crystals
Sigma-Aldrich
Phthaldialdehyde, ≥97% (HPLC), powder or crystals
Sigma-Aldrich
Phthaldialdehyde, suitable for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
Phthaldialdehyde Reagent, Solution Incomplete
Sigma-Aldrich
Phthaldialdehyde Reagent, Solution Complete