Merck
  • Estimation of relative reaction rate of hydroxy radical with poly-hydroxy benzenes: ESR spin trapping combined with UV-A photolysis.
Estimation of relative reaction rate of hydroxy radical with poly-hydroxy benzenes: ESR spin trapping combined with UV-A photolysis.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2013-03-12)
Seiko Nakagawa
ABSTRACT
The reaction of the hydroxy radical with thymidine and poly-hydroxy benzenes; i.e., catechol, resorcinol, hydroquinone, and pyrogallol, was studied by ESR spin trapping combined with the UV-A photolysis of hydrogen peroxide. The obtained relative reaction rates reasonably agreed with those previously obtained by pulse radiolysis. Electron distribution in the HOMO obtained by DFT calculations also supports the resulting order of reactivity of the polyphenols.
Sigma-Aldrich
Benzene, anhydrous, 99.8%
Sigma-Aldrich
Benzene, suitable for HPLC, ≥99.9%
Supelco
Benzene, analytical standard
Supelco
Benzene solution, certified reference material, TraceCERT®, 200 μg/mL in methanol
Sigma-Aldrich
Benzene, ACS reagent, ≥99.0%
Sigma-Aldrich
Benzene, puriss. p.a., reag. Ph. Eur., ≥99.7%
Supelco
Benzene, Pharmaceutical Secondary Standard; Certified Reference Material