Merck
[New approaches in the treatment of deep vein thrombosis].
Medicina clinica (2013-03-27)
José Antonio Nieto Rodríguez
ABSTRACT
In the last few years, several phase III clinical trials of new drug treatments for deep vein thrombosis (DVT) have been carried out or are about to finish. These drugs have a predictable and reliable pharmacokinetic and pharmacodynamic response and do not require monitoring and are consequently an attractive alternative for the treatment of a high proportion of patients with DVT. Dabigatran, edoxaban and idrabiotaparinux have been developed as an alternative to warfarin, and apixaban and rivaroxaban as one-drug only treatment for this disease, with a 1- or 3-week intensified phase of initial treatment, respectively. So far, the reported results show non-inferior efficacy and safety to warfarin or to conventional treatment. Therefore, the new anticoagulants will be particularly useful in patients with unstable INRs, warfarin incompatible pharmacologic interactions, and in those without access to regular coagulation monitoring.
Sigma-Aldrich
Biotin, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Biotin, tested according to Ph. Eur.
Sigma-Aldrich
Biotin, ≥99.0% (T)
Sigma-Aldrich
Biotin, meets USP testing specifications
Supelco
Biotin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Biotin, Pharmaceutical Secondary Standard; Certified Reference Material
Biotin, European Pharmacopoeia (EP) Reference Standard