Merck
  • Influence of material transition and interfacial area changes on flow and concentration in electro-osmotic flows.
Influence of material transition and interfacial area changes on flow and concentration in electro-osmotic flows.
Analytica chimica acta (2013-03-19)
Sudheer D Rani, Byoung-Hee You, Steve A Soper, Michael C Murphy, Dimitris E Nikitopoulos
ABSTRACT
This paper presents a numerical study to investigate the effect of geometrical and material transition on the flow and progression of a sample plug in electrokinetic flows. Three cases were investigated: (a) effect of sudden cross-sectional area change (geometrical transition or mismatch) at the interface, (b) effect of only material transition (i.e. varying ζ-potential), and (c) effect of combined material transition and cross-sectional area change at the interface. The geometric transition was quantified based on the ratio of reduced flow area A2 at the mismatch plane to the original cross-sectional area A1. Multiple simulations were performed for varying degrees of area reduction i.e. 0-75% reduction in the available flow area, and the effect of dispersion on the sample plug was quantified by standard metrics. Simulations showed that a 13% combined material and geometrical transition can be tolerated without significant loss of sample resolution. A 6.54% reduction in the flow rates was found between 0% and 75% combined material and geometrical transition.
Product Description
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 4,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), average Mw 97,000 (Typical), average Mn 46,000 (Typical)
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~15,000 by GPC, powder
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 8,000
Supelco
Poly(methyl methacrylate), analytical standard, 2,480,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 2,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 50,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 20,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 10,000
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~350,000 by GPC
Sigma-Aldrich
Poly(methyl methacrylate)
Sigma-Aldrich
Poly(methyl methacrylate)
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 100,000
Sigma-Aldrich
Poly(methyl methacrylate), isotactic, >80% isotactic