Merck
  • Synthesis of threose nucleic acid (TNA) triphosphates and oligonucleotides by polymerase-mediated primer extension.
Synthesis of threose nucleic acid (TNA) triphosphates and oligonucleotides by polymerase-mediated primer extension.
Current protocols in nucleic acid chemistry (2013-03-21)
Su Zhang, Hanyang Yu, John C Chaput
ABSTRACT
This unit describes the chemical synthesis of α-L-threofuranosyl nucleic acid (TNA) triphosphates for thymidine (T), guanosine (G), cytidine (C), and the diaminopurine (D) analog of adenosine and their incorporation into TNA oligonucleotides by enzyme-mediated polymerization of a DNA primer-template complex. Starting from suitably protected threofuranosyl nucleosides, TNA triphosphates are synthesized in a single-pot reaction and purified by ion-exchange and HPLC chromatography. Purified TNA triphosphates are diluted into stock solutions and used as substrates for the synthesis of TNA oligonucleotides. Oligonucleotide synthesis is accomplished using Therminator DNA polymerase, a commercial variant of the 9(o)N DNA polymerase bearing the A485L mutation.
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)
Sigma-Aldrich
Cytidine, 99%
Sigma-Aldrich
Thymidine, ≥99%
Sigma-Aldrich
Cytidine, BioReagent, suitable for cell culture, powder, ≥99%
Sigma-Aldrich
Sodium triphosphate pentabasic, purum p.a., ≥98.0% (T)
Sigma-Aldrich
Sodium tripolyphosphate, technical grade, 85%
Sigma-Aldrich
Guanosine, BioReagent, suitable for cell culture
Sigma-Aldrich
Guanosine, ≥98%
Sigma-Aldrich
Guanosine, ≥97.0% (HPLC)