Merck
The neurochemistry of music.
Trends in cognitive sciences (2013-04-02)
Mona Lisa Chanda, Daniel J Levitin
ABSTRACT
Music is used to regulate mood and arousal in everyday life and to promote physical and psychological health and well-being in clinical settings. However, scientific inquiry into the neurochemical effects of music is still in its infancy. In this review, we evaluate the evidence that music improves health and well-being through the engagement of neurochemical systems for (i) reward, motivation, and pleasure; (ii) stress and arousal; (iii) immunity; and (iv) social affiliation. We discuss the limitations of these studies and outline novel approaches for integration of conceptual and technological advances from the fields of music cognition and social neuroscience into studies of the neurochemistry of music.
Sigma-Aldrich
Corticotropin Releasing Factor sheep, ≥95% (HPLC)
Sigma-Aldrich
Oxytocin acetate salt hydrate, ≥97% (HPLC)
Oxytocin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Oxytocin, lyophilized powder, ~15 IU/mg solid (Prepared from synthetic oxytocin)
Sigma-Aldrich
Oxytocin