Merck
  • Optimization of single-cycle terahertz generation in LiNbO3 for sub-50 femtosecond pump pulses.
Optimization of single-cycle terahertz generation in LiNbO3 for sub-50 femtosecond pump pulses.
Optics express (2013-04-03)
Maksim Kunitski, Martin Richter, Mark D Thomson, Arno Vredenborg, Jian Wu, Till Jahnke, Markus Schöffler, Horst Schmidt-Böcking, Hartmut G Roskos, Reinhard Dörner
ABSTRACT
We compare different tilted-pulse-front pumping schemes for single-cycle THz generation in LiNbO(3) crystals both theoretically and experimentally in terms of conversion efficiency. The conventional setup with a single lens as an imaging element has been found to be highly inefficient in the case of sub-50 fs pump pulses, mainly due to the resulting chromatic aberrations. These aberrations are avoided in the proposed new setup, which employs two concave mirrors in a Keplerian telescope arrangement as the imaging sequence. This partially compensates spherical aberrations and results in a ca. six times higher conversion efficiency in the case of 35-fs optical pump pulse duration compared to the single-lens setup. A THz field strength of 60 kV/cm was obtained using 0.5 mJ pump pulses. The divergence of the THz beam has been found experimentally to depend on the pump imaging scheme employed.
Product Description
Niobium, IRMM®, certified reference material, 0.5 mm wire
Niobium, IRMM®, certified reference material, 0.5 mm wire
Niobium, IRMM®, certified reference material, 0.02 mm foil
Niobium, IRMM®, certified reference material, 0.02 mm foil
Sigma-Aldrich
Niobium, powder, <45 μm, 99.8% trace metals basis
Sigma-Aldrich
Niobium, foil, thickness 0.25 mm, 99.8% trace metals basis
Sigma-Aldrich
Lithium niobate, 99.9% trace metals basis