Merck
  • Sealing ability of various restorative materials as coronal barriers between endodontic appointments.
Sealing ability of various restorative materials as coronal barriers between endodontic appointments.
The journal of contemporary dental practice (2013-04-13)
Zalilah Tapsir, Hany Mohamed Aly Ahmed, Norhayati Luddin, Adam Husein
ABSTRACT
To evaluate and compare the microleakage of various restorative materials used as coronal barriers between endodontic appointments. Eighty extracted human permanent posterior teeth were prepared for standardized access cavities with dimensions of 4 × 4 × 4 mm. The teeth were then randomly divided into four groups; Kalzinol, Caviton, GC Fuji IX and GC Fuji II LC. After incubation, the samples were immersed in 2% methylene blue for 7 days. The depth of penetration was measured using a digital macroscope after longitudinal sectioning of each tooth. Kruskal-Wallis (p < 0.05) and multiple Mann-Whitney test with Bonferroni correction (p < 0.008) were used for data analysis. The degree of microleakage varied at the material/ tooth interface among the test materials, and the difference was statistically significant (p < 0.05). GC Fuji II LC group showed the least median microleakage value (0.8105 ± 0.305), followed by Caviton (1.1885 ± 0.396), GC Fuji IX (3.3985 ± 0.305) and Kalzinol (4.161 ± 0.853). Within the limitations of this study, GC Fuji II LC exhibited the best marginal seal, and has the potential to be used as a suitable coronal barrier between endodontic appointments. Given the prime importance that dental practitioners should thoroughly restore any tooth with a suitable coronal barrier between endodontic appointments, this study shows that Fuji II LC has the ability to maintain a hermetic seal for 7 days.
Product Description
Supelco
Eugenol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Drierite, with indicator, 10-20 mesh
Sigma-Aldrich
Drierite, with indicator, 6 mesh
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Eugenol, ReagentPlus®, 99%
Sigma-Aldrich
Eugenol, ≥98%, FCC, FG
Sigma-Aldrich
Eugenol, natural, ≥98%, FG
Supelco
Eugenol, PESTANAL®, analytical standard
Sigma-Aldrich
Drierite, with indicator, 4 mesh
Sigma-Aldrich
Drierite, with indicator, 8 mesh
Sigma-Aldrich
Poly(acrylic acid, sodium salt) solution, 45 wt. % in H2O
Sigma-Aldrich
Poly(acrylic acid, sodium salt) solution, average Mw ~8,000, 45 wt. % in H2O
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Supelco
Poly(acrylic acid sodium salt), analytical standard, suitable for gel permeation chromatography (GPC), 1,250
Sigma-Aldrich
Calcium sulfate, −325 mesh, 99%
Sigma-Aldrich
Calcium sulfate dihydrate, ACS reagent, 98%
Sigma-Aldrich
Calcium sulfate dihydrate, puriss., meets analytical specification of NF, E 516, 99.0-101.0% (based on anhydrous substance)
Sigma-Aldrich
Calcium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium sulfate dihydrate, ReagentPlus®, ≥99%
Sigma-Aldrich
Methylene Blue solution, suitable for microscopy
Sigma-Aldrich
Methylene Blue solution, concentrate according to Ehrlich, concentrated, aqueous solution
Sigma-Aldrich
Methylene Blue solution, suitable for microbiology
Sigma-Aldrich
Poly(acrylic acid sodium salt), average Mw ~2,100
Sigma-Aldrich
Poly(acrylic acid sodium salt), average Mw ~5,100 by GPC, powder
Supelco
Poly(acrylic acid sodium salt), analytical standard, suitable for gel permeation chromatography (GPC), 16,000