Merck
  • Modulation of cytotoxicity and metabolism of 5-fluorouracil in two intestine cell lines.
Modulation of cytotoxicity and metabolism of 5-fluorouracil in two intestine cell lines.
Advances in experimental medicine and biology (1986-01-01)
G J Peters, E Laurensse, A Leyva, H M Pinedo