Merck
  • Quality control slide for potassium hydroxide and cellufluor fungal preparations.
Quality control slide for potassium hydroxide and cellufluor fungal preparations.
Journal of clinical microbiology (1989-06-01)
J L Perry, G R Miller
ABSTRACT
An opaque, water-insoluble quality control material with a skinlike microscopic appearance was prepared by inoculating melted xanthine (0.4%) agar with filamentous fungi and dispensing drops onto glass slides. After solidification of the agar, the material was rapidly cleared by 10% KOH, revealing fungal elements stained by Cellufluor reagent.