Merck
  • Inhibition of beta-adrenergic stimulated calcium pump of rat cardiac sarcoplasmic reticulum by tricyclohexyltin hydroxide.
Inhibition of beta-adrenergic stimulated calcium pump of rat cardiac sarcoplasmic reticulum by tricyclohexyltin hydroxide.
Cell biochemistry and function (1987-04-01)
I K Sahib, D Desaiah
ABSTRACT
The effects of tricyclohexyltin hydroxide (Plictran), an organotin acaricide, on 45Ca2+ uptake and Ca2+ ATPase were studied in vitro and in vivo in rat heart ventricular membrane vesicles, primarily sarcoplasmic reticulum. There was a concentration dependent inhibition of both 45Ca2+ uptake and Ca2+ ATPase in vivo as well as in vitro. Isoproterenol, a beta-adrenergic agonist, stimulated 45Ca2+ uptake and Ca2+ ATPase of sarcoplasmic reticulum and this was also inhibited by Plictran. Since cardiac relaxation is mediated by beta-adrenergic stimulation via Ca+ uptake by sarcoplasmic reticulum, the inhibition of calcium pump activity by Plictran may result in alterations in cardiac Ca2+ fluxes leading to cardiac dysfunction.