Merck
  • Hemolytic activity of a cyclic peptide Ro09-0198 isolated from Streptoverticillium.
Hemolytic activity of a cyclic peptide Ro09-0198 isolated from Streptoverticillium.
Biochimica et biophysica acta (1988-05-24)
S Y Choung, T Kobayashi, J Inoue, K Takemoto, H Ishitsuka, K Inoue
ABSTRACT
Ro09-0198, a cyclic peptide isolated from culture filtrates of Streptoverticillium griseoverticillatum, induced lysis of erythrocytes. Preincubation of the peptide with phosphatidylethanolamine reduced the hemolytic activity, whereas other phospholipids present in erythrocytes in nature had no effect. A study of the structural requirements on phosphatidylethanolamine necessary for interaction with the peptide indicates that Ro09-0198 recognizes strictly a particular chemical structure of phosphatidylethanolamine: dialkylphosphoethanolamine as well as 1-acylglycerophosphoethanolamine showed the same inhibitory effect on hemolysis induced by Ro09-0198 as diacylphosphatidylethanolamine, whereas phosphoethanolamine gave no inhibitory effect. Neither phosphatidyl-N-monomethylethanolamine nor alkylphosphopropanolamine had an inhibitory effect. Consequently, the hydrophobic chain is necessary for the interaction and the phosphoethanolamine moiety is exactly recognized by the peptide. Ro-09-0198-induced hemolysis was temperature-dependent and the sensitivity of hemolysis differed greatly among animal species.
Sigma-Aldrich
Cinnamycin, from Streptomyces cinnamoneus, ≥95% (HPLC)