- Sterol biosynthesis via cycloartenol and other biochemical features related to photosynthetic phyla in the amoeba Naegleria lovaniensis and Naegleria gruberi.
Sterol biosynthesis via cycloartenol and other biochemical features related to photosynthetic phyla in the amoeba Naegleria lovaniensis and Naegleria gruberi.
The sterols and sterol precursors of two amoebae of the genus Naegleria, Naegleria lovaniensis and Naegleria gruberi were investigated. Cycloartenol, the sterol precursor in photosynthetic organisms, is present in both amoebae. In N. lovaniesis, it is accompanied by lanosterol and parkeol, as well as by the 24,25-dihydro derivatives of these triterpenes. One of the most striking features of these amoebae is the accumulation of 4 alpha-methylsterols which are present in similar amounts as those of 4,4-desmethylsterols (3-5 mg/g, dry weight). 4 alpha-Methylergosta-7,22-dienol was identified as a new compound. Ergosterol was the major 4,4-desmethylsterol, accompanied by small amounts of C27 and other C28 sterols. Treatment of N. lovaniensis with fenpropimorph modified the sterol pattern of this amoeba and inhibited its growth. This fungicide, known to inhibit steps of sterol biosynthesis in fungi and plants, induced the disappearance of 4 alpha-methyl-delta 7-sterols and the appearance of the unusual delta 6,8,22-ergostatrienol as in A. polyphaga. These results might be explained by a partial inhibition of the delta 8----delta 7 isomerase, the small amounts of delta 7-sterols formed being converted into ergosterol which is still present in fenpropimorph-exposed cells. De novo sterol biosynthesis in N. lovaniensis was shown by incorporation of [1-14C]acetate into sterols and sterol precursors, especially cycloartenol. Lanosterol and parkeol were not significantly labelled. Furthermore, [3-3H]squalene epoxide was efficiently cyclized by a cell-free system of this amoeba into cycloartenol, and again no significant radioactivity was detected in lanosterol and parkeol. This shows that cycloartenol, the sterol precursor in plants and algae, is also the sterol precursor in Naegleria species, and that these amoebae, like A. polyphaga, are related by some biosynthetic pathways to photosynthetic phyla. Lanosterol, the sterol precursor in non-photosynthetic phyla (animal and fungi) and parkeol are more likely dead-ends of this biosynthetic pathway. The peculiar phylogenetic position of these protozoa was further emphasized by the action of indole acetic acid and other auxine-like compounds on their growth. Indeed amoebic growth was enhanced in the presence of these higher plant growth hormones. The differences in the sterol composition of the protozoa we have hitherto examined is related to their sensitivity toward polyene macrolide antibiotics.(ABSTRACT TRUNCATED AT 400 WORDS)