Merck
  • Clinical, biochemical, and hormonal aspects of treatment with Des-tyr1-gamma-endorphin in schizophrenia.
Clinical, biochemical, and hormonal aspects of treatment with Des-tyr1-gamma-endorphin in schizophrenia.
Psychiatry research (1984-04-01)
W M Verhoeven, J M van Ree, H G Westenberg, J M Krul, G J Brouwer, J H Thijssen, D de Wied, H M van Praag, D L Ceulemans, R S Kahn
ABSTRACT
Des-tyr1-gamma-endorphin (DT gamma E) was administered intramuscularly in a dose of 1 mg/day for 10 days to 18 neuroleptic-free schizophrenic patients in a double-blind crossover design. Six patients showed either a slight or no antipsychotic response; seven patients showed a moderate antipsychotic response; and the remaining five patients showed a marked antipsychotic response. DT gamma E led to a decrease of plasma prolactin levels in patients treated with DT gamma E in the first period of experimental treatment as compared to those treated with placebo. Neither plasma levels of growth hormone and cortisol nor cerebrospinal fluid concentrations of homovanillic acid, 5-hydroxyindoleacetic acid, and 3-methoxy-4-hydroxyphenylglycol were affected by DT gamma E. Patients suffering from a hebephrenic or paranoid type of schizophrenia and those presenting relatively fewer negative symptoms were most susceptible to treatment with DT gamma E. These data confirm and extend previous findings that DT gamma E has antipsychotic properties in a number of schizophrenic patients.