Merck
  • Comparative mutagenicity of two triarylmethane food dyes in Salmonella, Saccharomyces and Drosophila.
Comparative mutagenicity of two triarylmethane food dyes in Salmonella, Saccharomyces and Drosophila.
Food and cosmetics toxicology (1981-08-01)
D S Angus, R S Baker, A M Bonin, D Callen, A M Clark
Sigma-Aldrich
Coomassie Violet R200, Dye content 50 %