Merck
  • Anxiolytics' effects on the actual driving performance of patients and healthy volunteers in a standardized test. An integration of three studies.
Anxiolytics' effects on the actual driving performance of patients and healthy volunteers in a standardized test. An integration of three studies.
Neuropsychobiology (1995-01-01)
J F O'Hanlon, A Vermeeren, M M Uiterwijk, L M van Veggel, H F Swijgman
ABSTRACT
Effects of benzodiazepine (diazepam, lorazepam) and benzodiazepine-like anxiolytics (alpidem, suriclone) and a 5-HT-3 antagonist (ondansetron) on actual driving performance were measured in three double-blind, placebo-controlled studies. Subjects were healthy volunteers in two and anxious patients in the third. Treatments lasted for 8 days. Standardized testing occurred within the first full day and on the last day of treatment. No important differences existed between volunteers' and patients' baseline and/or placebo performances and both groups responded similarly to comparable drugs/doses. Benzodiazepine and benzodiazepine-like anxiolytics produced marked and pervasive driving impairment, which lasted throughout treatment; but ondansetron, none.