Merck
  • Vagal release of vasoactive intestinal peptide can promote vagotonic tachycardia in the isolated innervated rat heart.
Vagal release of vasoactive intestinal peptide can promote vagotonic tachycardia in the isolated innervated rat heart.
Cardiovascular research (1994-12-01)
A Shvilkin, P Danilo, P Chevalier, F Chang, I S Cohen, M R Rosen
ABSTRACT
The aim was to determine the extent to which endogenous release of vasoactive intestinal polypeptide (VIP) might be implicated in the modulation of sinoatrial rate in the presence and absence of muscarinic blockade or beta blockade. Langendorff perfused rat hearts were studied with the right vagus intact. The hearts were maintained in sinus rhythm and subjected to right vagal stimuli of 5, 10, 20, and 30 Hz. Administration of exogenous VIP, 10(-8) M, increased sinus rate by 20% (p < 0.05). This increase in heart rate was reduced significantly to 8% by the VIP antagonist [D-p-Cl-Phe6, Leu17]VIP, 10(-7) M, which alone had no effect on sinus rate. Vagal stimulation reduced sinus rate from a control of 254(SEM 2) to 164(17) beats.min-1 (p < 0.05) at 20 Hz. VIP, 10(-8) M, increased these rates to 284(6) and 220(21) beats.min-1 (p < 0.05). In another eight vagally stimulated hearts, frequencies of 5-20 Hz reduced sinus rate. At 30 Hz heart rate increased in five, and the resultant rate was significantly faster in these [154(10) beats.min-1] than in the remainder [98(12) beats.min-1, p < 0.05]. Vagal stimulation also increased sinus rate (p < 0.05) in four of seven additional hearts perfused with atropine, 2 x 10(-6) M. This increase was completely abolished by [D-p-Cl-Phe6, Leu17]VIP. That the effect was not beta adrenergic was demonstrated in eight experiments using atropine plus propranolol, 1 x 10(-7) M. A vagally induced increment in rate still occurred (p < 0.05) and was abolished by [D-p-CL-Phe6, Leu17]VIP. The ability to ascribe a rate change to VIP release was maximal in the presence of propranolol and atropine, intermediate in the presence of atropine alone, and minimal in the absence of muscarinic or beta blockade. Vagally released VIP is capable of limiting the decrement in sinus rate that occurs at high frequencies of vagal stimulation, and in some circumstances can actually increment sinus rate. Its role as an endogenous modulator of vagal effects on heart rate and as a possible cause of vagal and postvagal tachycardias should be further explored.