Merck
  • Hair analysis for drugs of abuse. VI. The excretion of methoxyphenamine and methamphetamine into beards of human subjects.
Hair analysis for drugs of abuse. VI. The excretion of methoxyphenamine and methamphetamine into beards of human subjects.
Forensic science international (1993-12-01)
Y Nakahara, K Takahashi, K Konuma
ABSTRACT
The excretion of methoxyphenamine (MOP) and methamphetamine (MA) into beards has been studied. Six healthy male subjects orally took 50 mg of MOP at a single dose and 7 doses for a successive 7 days. Their beard hairs were collected by an electric shaver every morning until MOP disappeared from the beard. After washing with 0.1% SDS, the beard samples were extracted with methanol-5 N HCl (20:1) under ultra-sonication for 1 h and the solution was kept overnight. MOP in the extract was determined by GC/MS using deuterium labelled MOP as an internal standard after trifluoroacetyl-derivatization. The drug concentrations in beard and the reproducibility of analysis were compared with the three procedures, unwashed, 0.1% SDS (wash I) and the additional ethanol (wash II) wash. The drug concentration in beard after SDS wash was 0.5-2.5 ng/mg lower than that in unwashed beard during the first 5-6 days. The drug concentration in beard after ethanol wash was much lower than that in the unwashed beard. The drug excreted into beard was detected 10 approximately 12 days for a single dose and 12-14 days for 7 doses after the last dosage at the cut off level of 1 ng/mg. On the contrary, the drug excreted in urine was not detected after more than 3 days after use. O-Desmethyl MOP, a major metabolite of MOP, was also detected in beard. The procedures were applied to the detection of MA in beard of MA abusers. It was realized that a beard sample was more useful than a urine sample assuming a longer detection.
Sigma-Aldrich
Methoxyphenamine hydrochloride, ≥99%
Pricing and availability is not currently available.