Merck
  • Different biotransformation of morphine in isolated liver cells from guinea pig and rat.
Different biotransformation of morphine in isolated liver cells from guinea pig and rat.
Biochemical pharmacology (1993-09-14)
T A Aasmundstad, A Ripel, E Bodd, A Bjørneboe, J Mørland
ABSTRACT
The biotransformation of morphine was characterized in freshly isolated parenchymal and non-parenchymal liver cells from rats and guinea pigs in suspension culture to establish an in vitro model for morphine metabolism. Liver cells were prepared by a collagenase perfusion technique, and separated by differential centrifugation. Morphine metabolism was investigated at different concentrations (1, 5, 100 and 200 microM). Samples were taken repeatedly during 2-4 hr of incubation, and subsequently analysed on a HPLC system employing both UV and electrochemical detection. In suspensions of hepatocytes from both animal species morphine-3-glucuronide (M3G) was the major metabolite of morphine, and was formed at comparable rates at all concentrations examined. Guinea pig hepatocytes in addition produced considerable quantities of morphine-6-glucuronide (M6G), whereas this metabolite was detected only intracellularly in minor quantities in rat hepatocytes. The ratio between the two morphine glucuronides (M3G/M6G) in suspensions of guinea pig hepatocytes was approximately 4:1. N-Demethylation of morphine was more pronounced per mg cell protein in rat hepatocytes compared to guinea pig cells. Metabolic activity towards morphine was not detected in non-parenchymal cells of the two species. The morphine glucuronidation pattern found in guinea pig hepatocytes resembles to a greater extent than that found in rat hepatocytes the pattern found in in vivo studies of humans. It was concluded that isolated guinea pig parenchymal cells appeared to be a promising in vitro system for studies of morphine glucuronidation, and to observe metabolism in general.
Product Description
Supelco
Normorphine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®