Merck
  • Carboxydipeptidase from Boophilus microplus: a "concealed" antigen with similarity to angiotensin-converting enzyme.
Carboxydipeptidase from Boophilus microplus: a "concealed" antigen with similarity to angiotensin-converting enzyme.
Insect biochemistry and molecular biology (1995-10-01)
J M Jarmey, G A Riding, R D Pearson, R V McKenna, P Willadsen
ABSTRACT
A protein, Bm91, which was first identified as a protective vaccine antigen from the tick Boophilus microplus, has regions of very strong amino acid sequence similarity to mammalian carboxydipeptidases or angiotensin converting enzymes (ACE; E.C. 3.4.15.1). This protein is now shown to share many biochemical and enzymatic properties with mammalian carboxydipeptidases. It is enzymatically active in a conventional assay for ACE using hippuryl-Gly-Gly as substrate. The hydrolysis of the C-terminal nonapeptide of the insulin B chain proceeds by sequential removal of carboxy-terminal dipeptides. The similarities extend to the dependence of activity on pH and added salt. Bm91 is inhibited by two well-characterized inhibitors of the mammalian enzymes, the drug Captopril and a nonapeptide, and the inhibition occurs in similar concentration ranges to those effective with the mammalian enzymes. However, the natural substrates of the tick enzyme are unknown. Angiotensin I itself is a poor substrate and the enzyme's natural substrates are likely to be one or more of the pharmacologically active peptides occurring in insects and arthropods.