Merck
  • Solid-phase extraction and HPLC analysis of kebuzone and its metabolites in blood.
Solid-phase extraction and HPLC analysis of kebuzone and its metabolites in blood.
Die Pharmazie (1996-09-01)
J Klimes, J Sochor, J Sedlacek
ABSTRACT
A HPLC method for quantification of kebuzone and its metabolites in whole blood was developed. The compounds and the internal standard were isolated from blood by solid-phase extraction on a C-18 cartridge. A blood sample was to be hemolyzed before extraction. HPLC was performed on a C-18 column with the mobile phase composed of methanol/water acidified to pH 2.7 and UV absorbance detection at 247 nm. This method has been successfully applied to a pharmacokinetic study of kebuzone and its metabolites in rabbits.
Product Description
Supelco
Methaqualone solution, 1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®