Merck
  • The activities of four anticancer alkyllysophospholipids against Leishmania donovani, Trypanosoma cruzi and Trypanosoma brucei.
The activities of four anticancer alkyllysophospholipids against Leishmania donovani, Trypanosoma cruzi and Trypanosoma brucei.
The Journal of antimicrobial chemotherapy (1996-12-01)
S L Croft, D Snowdon, V Yardley
ABSTRACT
The in-vitro activities of four anticancer alkyllysophospholipids, ET-18-OCH3 (edelfosine), hexadecylphosphocholine (miltefosine), ilmofosine and SRI 62-834, were determined with ED50 values for Leishmania donovani and Trypanosoma cruzi amastigotes between 0.2 and 5.0 microM and for Trypanosoma brucei trypomastigotes between 7.0 and 50.8 microM. In BALB/c mice miltefosine and ilmofosine were the most active compounds with ED50 values of 2.9 and 14.5 mg/kg x 5 doses against L. donovani and a suppressive effect on T. cruzi infections at 30 mg/kg x 5 doses.