Merck
  • A fatty acid synthase gene in Cochliobolus carbonum required for production of HC-toxin, cyclo(D-prolyl-L-alanyl-D-alanyl-L-2-amino-9, 10-epoxi-8-oxodecanoyl).
A fatty acid synthase gene in Cochliobolus carbonum required for production of HC-toxin, cyclo(D-prolyl-L-alanyl-D-alanyl-L-2-amino-9, 10-epoxi-8-oxodecanoyl).
Molecular plant-microbe interactions : MPMI (1997-03-01)
J H Ahn, J D Walton
ABSTRACT
The fungal maize pathogen Cochliobolus carbonum produces a phytotoxic and cytostatic cyclic peptide, HC-toxin, of structure cyclo(D-prolyl-L-alanyl-D-alanyl-L-Aeo), in which Aeo stands for 2-amino-9,10-epoxi-8-oxodecanoic acid. Here we report the isolation of a gene, TOXC, that is present only in HC-toxin-producing (Tox2+) fungal strains. TOXC is present in most Tox2+ strains in three functional copies, all of which are on the same chromosome as the gene encoding HC-toxin synthetase. When all copies of TOXC are mutated by targeted gene disruption, the fungus grows and sporulates normally in vitro but no longer makes HC-toxin and is not pathogenic, indicating that TOXC has a specific role in HC-toxin production and hence virulence. The TOXC mRNA is 6.5 kb and the predicted product has 2,080 amino acids and a molecular weight of 233,000. The primary amino acid sequence is highly similar (45 to 47% identity) to the beta subunit of fatty acid synthase from several lower eukaryotes, and contains, in the same order as in other beta subunits, domains predicted to encode acetyl transferase, enoyl reductase, dehydratase, and malonyl-palmityl transferase. The most plausible function of TOXC is to contribute to the synthesis of the decanoic acid backbone of Aeo.