Merck
Gas chromatography-mass spectrometry of Alternaria mycotoxins.
Journal of chromatography. A (1997-03-28)
P M Scott, D Weber, S R Kanhere
ABSTRACT
Heptafluorobutyrate (HFB) derivatives have not previously been used for GC of Alternaria mycotoxins. Capillary (0.5 micron film) GC-mass spectrometry (MS) showed that full and partial derivatives of alternariol (AOH), alternariol monomethyl ether (AME) and altenuene (ALT); a structurally uncharacterized derivative of altertoxin I (ALTX-I); and a tris-HFB derivative of tenuazonic acid (TA) were formed with heptafluorobutyric anhydride and a basic catalyst. Full and partial trimethylsilyl (TMS) ethers of these mycotoxins were formed with Tri-Sil TBT. Apple juice extracts caused increased response in GC-MS of AOH bis-HFB and bis-TMS derivatives. Natural occurrence of AOH in apple juice has been demonstrated.