Merck
  • Biosynthesis of a cyclic tautomer of (3-methylmaleyl)acetone from 4-hydroxy-3,5-dimethylbenzoate by Pseudomonas sp. HH35 but not by Rhodococcus rhodochrous N75.
Biosynthesis of a cyclic tautomer of (3-methylmaleyl)acetone from 4-hydroxy-3,5-dimethylbenzoate by Pseudomonas sp. HH35 but not by Rhodococcus rhodochrous N75.
Biochemical and biophysical research communications (1997-09-23)
R B Cain, P Fortnagel, S Hebenbrock, G W Kirby, H J McLenaghan, G V Rao, S Schmidt
ABSTRACT
Here we report that the bacterial catabolism of 4-hydroxy-3,5-dimethylbenzoic acid 1 takes a different course in Rhodococcus rhodochrous N75 and Pseudomonas sp. strain HH35. The former organism accumulates a degradation metabolite of the acid which we isolated and identified as 2,6-dimethylhydroquinone 2. The latter bacterial strain converts the acid and the hydroquinone into a dead-end metabolite. This novel compound was characterised unequivocally by mass spectrometry and 1H and 18C NMR and UV spectroscopy as 4-acetonyl-4-hydroxy-2-methylbut-2-en-1,4-olide 4, a cyclic tautomer of (3-methylmaleyl)acetone, which exists as the enol carboxylate form 8 in aqueous solution.
Sigma-Aldrich
3,5-Dimethylbenzoic acid, 99%
Pricing and availability is not currently available.