Merck
  • Involvement of thromboxane A2 in the mediation of the contractile effect induced by inhibition of nitric oxide synthesis in isolated rat middle cerebral arteries.
Involvement of thromboxane A2 in the mediation of the contractile effect induced by inhibition of nitric oxide synthesis in isolated rat middle cerebral arteries.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism (1998-06-17)
Z Benyó, C Görlach, M Wahl
ABSTRACT
Inhibition of nitric oxide (NO) synthesis induces vasoconstriction and reduction of the blood flow in the brain, indicating that basal release of NO provides a resting vasorelaxant tone in the cerebral circulation. In the present study, the contractile effect of the NO synthase blocker NG-nitro-L-arginine (100 mumol/L) in isolated rat middle cerebral arteries was attenuated markedly in the presence of the cyclooxygenase inhibitor indomethacin (5 mumol/L), the thromboxane A2 synthase inhibitor ridogrel (10 mumol/L), or the thromboxane receptor antagonist ICI 192605 (100 mumol/L). These results indicate that removal of the endogenous NO stimulates the release of thromboxane A2 in cerebral vessels and basal NO production regulates the resting cerebrovascular tone, at least in part, by suppressing thromboxane A2.
Sigma-Aldrich
ICI 192605, ≥98% (HPLC)
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)