Merck
  • A key role for CC chemokine receptor 4 in lipopolysaccharide-induced endotoxic shock.
A key role for CC chemokine receptor 4 in lipopolysaccharide-induced endotoxic shock.
The Journal of experimental medicine (2000-05-17)
Y Chvatchko, A J Hoogewerf, A Meyer, S Alouani, P Juillard, R Buser, F Conquet, A E Proudfoot, T N Wells, C A Power
ABSTRACT
CC chemokine receptor (CCR)4, a high affinity receptor for the CC chemokines thymus and activation-regulated chemokine (TARC) and macrophage-derived chemokine (MDC), is expressed in the thymus and spleen, and also by peripheral blood T cells, macrophages, platelets, and basophils. Recent studies have shown that CCR4 is the major chemokine receptor expressed by T helper type 2 (Th2) polarized cells. To study the in vivo role of CCR4, we have generated CCR4-deficient (CCR4(-/-)) mice by gene targeting. CCR4(-/-) mice developed normally. Splenocytes and thymocytes isolated from the CCR4(-/-) mice failed to respond to the CCR4 ligands TARC and MDC, as expected, but also surprisingly did not undergo chemotaxis in vitro in response to macrophage inflammatory protein (MIP)-1alpha. The CCR4 deletion had no effect on Th2 differentiation in vitro or in a Th2-dependent model of allergic airway inflammation. However, CCR4(-/-) mice exhibited significantly decreased mortality on administration of high or low dose bacterial lipopolysaccharide (LPS) compared with CCR4(+/+) mice. After high dose LPS treatment, serum levels of tumor necrosis factor alpha, interleukin 1beta, and MIP-1alpha were reduced in CCR4(-/-) mice, and decreased expression of MDC and MIP-2 mRNA was detected in peritoneal exudate cells. Analysis of peritoneal lavage cells from CCR4(-/)- mice by flow cytometry also revealed a significant decrease in the F4/80(+) cell population. This may reflect a defect in the ability of the CCR4(-/-) macrophages to be retained in the peritoneal cavity. Taken together, our data reveal an unexpected role for CCR4 in the inflammatory response leading to LPS-induced lethality.