Merck
  • An autopsy case of poisoning by massive absorption of cresol a short time before death.
An autopsy case of poisoning by massive absorption of cresol a short time before death.
Forensic science international (2002-04-17)
Jun Monma-Ohtaki, Yoshitaka Maeno, Masataka Nagao, Mineo Iwasa, Hiroyoshi Koyama, Ichiro Isobe, Yoshimi Seko-Nakamura, Tsukasa Tsuchimochi, Tomohiro Matsumoto
ABSTRACT
A 65-year-old male patient who was hospitalized with schizophrenia died about 15 min later after ingestion of a large volume of saponated cresol solution in a mental hospital. Fatal levels of free p- and m-cresol in the heart blood were detected at 458.8 and 957.3 microg/ml, respectively, which far exceeded the fatal levels determined previously. The levels in the heart muscle, liver and spleen tissues were also extremely high, and there was 250 ml of cresol-odor-emitting fluid in the stomach. The levels of glucuronic-acid-conjugated p- and m-cresols in the heart blood were 38.2 and 85.6 microg/ml, respectively. Although the high levels of cresols in the heart blood may be due to diffusion from the stomach contents, it is surmised that the essential levels of free and conjugated forms in blood were at least 99 and 240 microg/ml, respectively, considering the results of postmortem examinations and some case reports. It was concluded that about 340 microg/ml of the total cresols was absorbed in a very short period following oral ingestion of saponated cresol solution in this case.
Sigma-Aldrich
m-Cresol, 99%
Supelco
m-Cresol, analytical standard
Sigma-Aldrich
m-Cresol, ≥98%, FG
Sigma-Aldrich
o-Cresol, ≥99%
Supelco
o-Cresol, analytical standard
Sigma-Aldrich
o-Cresol, ReagentPlus®, ≥99%