Merck
Plasma pteridine concentrations in patients with chronic renal failure.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association (2002-05-29)
Keitaro Yokoyama, Masamich Tajima, Hiraku Yoshida, Masaaki Nakayama, Goro Tokutome, Hiroshi Sakagami, Tatsuo Hosoya
ABSTRACT
Pteridine metabolism is impaired in the uraemic state. This may affect cardiovascular function and contribute to malnutrition. We wished to clarify further the impact of impaired pteridine metabolism. Using the HPLC method, the plasma concentrations of endogenous pteridines were determined in 64 patients with chronic renal failure (33 on intermittent haemodialysis (HD) treatment vs 31 not yet on renal replacement therapy), and in 18 healthy controls. The patients were classified into three groups on the basis of creatinine clearance (Ccr): group (a), Ccr >60 ml/min; group (b), Ccr=10-60 ml/min; group (c), all patients receiving HD. Total neopterin (NP) and biopterin (BP) levels and the NP/BP ratio (a biomarker for macrophage activity) were significantly higher, whereas tetrahydrobiopterin (BH(4))/dihydrobiopterin (BH(2)) ratio (a biomarker for nitric oxide synthase and phenylalanine hydroxylase activities) was significantly lower in group (c) (118.9+/-11.7 ng/ml, 18.8+/-1.2 ng/ml, 6.79+/-0.53, and 0.26+/-0.06) than in healthy subjects (5.17+/-0.29 ng/ml, 2.83+/-0.19 ng/ml, 1.92+/-0.13, and 1.15+/-0.11; P<0.01). These significant differences were also observed between control and group (b) (12.4+/-2.20 ng/ml, 4.48+/-0.36 ng/ml, 2.81+/-0.48, and 0.74+/-0.08; P<0.01). In groups (a) and (b), significant negative correlations were found between Ccr and the total NP level (r=-0.663, P<0.01), the total BP level (r=-0.492, P<0.01), the BH(2) level (r=-0.677, P<0.01), and the NP/BP ratio (r=-0.493, P<0.01). Conversely, significant positive correlations were found between Ccr and the BH(4)/BH(2) ratio (r=0.602, P<0.01). The reduction of quinoid-type BH(2) to BH(4) is modified in patients with advanced chronic renal failure, before and after the initiation of regular HD treatment. These metabolic alterations may play a role in the impaired macrophage, endothelial constitutive nitric oxide synthase, or phenylalanine hydroxylase (PH) activities observed in such patients.
Product Description
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Cadaverine, 95%
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Zinc preparation, 5 g/dL Zn2+ in THF, highly reactive Rieke®metal
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cadaverine, purum, ≥97.0% (GC)
Supelco
Acetone, analytical standard
Sigma-Aldrich
Trimethylamine solution, 31-35 wt. % in ethanol, 4.2 M, contains toluene as stabilizer
Sigma-Aldrich
Zinc, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, shot, <12 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, sticks, diam. 7-10 mm, 99.97% trace metals basis
Sigma-Aldrich
Zinc, shot, 5 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, pieces, 2-14 mesh, 99.9% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, dust, <10 μm, ≥98%
Sigma-Aldrich
Trimethylamine solution, 25 wt. % in H2O
Sigma-Aldrich
Trimethylamine solution, 43.0-49.0% in H2O (T)
Sigma-Aldrich
Zinc, granular, 20-30 mesh, ACS reagent, ≥99.8%
Sigma-Aldrich
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®