Merck
  • Increased plasma phenylacetic acid in patients with end-stage renal failure inhibits iNOS expression.
Increased plasma phenylacetic acid in patients with end-stage renal failure inhibits iNOS expression.
The Journal of clinical investigation (2003-07-17)
J Jankowski, M van der Giet, V Jankowski, S Schmidt, M Hemeier, B Mahn, G Giebing, M Tolle, H Luftmann, H Schluter, W Zidek, M Tepel
ABSTRACT
NO prevents atherogenesis and inflammation in vessel walls by inhibition of cell proliferation and cytokine-induced endothelial expression of adhesion molecules and proinflammatory cytokines. Reduced NO production due to inhibition of either eNOS or iNOS may therefore reinforce atherosclerosis. Patients with end-stage renal failure show markedly increased mortality due to atherosclerosis. In the present study we tested the hypothesis that uremic toxins are responsible for reduced iNOS expression. LPS-induced iNOS expression in mononuclear leukocytes was studied using real-time PCR. The iNOS expression was blocked by addition of plasma from patients with end-stage renal failure, whereas plasma from healthy controls had no effect. Hemofiltrate obtained from patients with end-stage renal failure was fractionated by chromatographic methods. The chromatographic procedures revealed a homogenous fraction that inhibits iNOS expression. Using gas chromatography/mass spectrometry, this inhibitor was identified as phenylacetic acid. Authentic phenylacetic acid inhibited iNOS expression in a dose-dependent manner. In healthy control subjects, plasma concentrations were below the detection level, whereas patients with end-stage renal failure had a phenylacetic acid concentration of 3.49 +/- 0.33 mmol/l (n = 41). It is concluded that accumulation of phenylacetic acid in patients with end-stage renal failure inhibits iNOS expression. That mechanism may contribute to increased atherosclerosis and cardiovascular morbidity in patients with end-stage renal failure.
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Zinc preparation, 5 g/dL Zn2+ in THF, highly reactive Rieke®metal
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-25, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Zinc, shot, <12 mm, 99.99% trace metals basis
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Supelco
Acetone, analytical standard
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Cadaverine, purum, ≥97.0% (GC)
Sigma-Aldrich
Cadaverine, 95%
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Zinc, dust, <10 μm, ≥98%
Sigma-Aldrich
Zinc, pieces, 2-14 mesh, 99.9% trace metals basis
Sigma-Aldrich
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Sigma-Aldrich
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Zinc, granular, 20-30 mesh, ACS reagent, ≥99.8%
Sigma-Aldrich
Trimethylamine solution, 31-35 wt. % in ethanol, 4.2 M, contains toluene as stabilizer
Sigma-Aldrich
Trimethylamine solution, 43.0-49.0% in H2O (T)
Sigma-Aldrich
Zinc, shot, 5 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, sticks, diam. 7-10 mm, 99.97% trace metals basis
Sigma-Aldrich
Trimethylamine solution, 25 wt. % in H2O
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Acetone, ≥99%, FCC, FG