Merck
  • Silicon chemistry as a novel source of chemical diversity in drug design.
Silicon chemistry as a novel source of chemical diversity in drug design.
Current opinion in drug discovery & development (2003-09-04)
William Bains, Reinhold Tacke
ABSTRACT
The use of organosilicon chemistry in drug design is reviewed. The field of bioorganosilicon chemistry, which sprung up in the 1970s to exploit the opportunities of silicon for drug design, is currently being developed into a practical and commercial enterprise. This review is mainly focused on two different approaches: (i) synthesizing a silicon analog of a known drug in which one carbon atom has been replaced by a silicon atom, with the rest of the molecule being identical (carbon/silicon switch, sila-substitution); (ii) synthesizing completely new silicon-based classes of compounds (the carbon analogs of which do not exist) that exploit specific features of silicon chemistry to address a well-validated target.
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, N-type, contains no dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, N-type, contains phosphorus as dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer, <111>, P-type, contains boron as dopant, diam. × thickness 2 in. × 0.3 mm
Sigma-Aldrich
Silicon, powder, −325 mesh, 99% trace metals basis
Sigma-Aldrich
Silicon, wafer (single side polished), <111>, N-type, contains no dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <111>, N-type, contains no dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, P-type, contains boron as dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <111>, P-type, contains boron as dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, N-type, contains phosphorus as dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, nanopowder, <100 nm particle size (TEM), ≥98% trace metals basis
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, N-type, contains no dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, P-type, contains boron as dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), contains phosphorus as dopant, <111>, N-type, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, pieces, 99.95% trace metals basis