Merck
  • Serum concentrations of asymmetric (ADMA) and symmetric (SDMA) dimethylarginine in patients with chronic kidney diseases.
Serum concentrations of asymmetric (ADMA) and symmetric (SDMA) dimethylarginine in patients with chronic kidney diseases.
Clinica chimica acta; international journal of clinical chemistry (2003-09-23)
Christian Fleck, Frank Schweitzer, Elke Karge, Martin Busch, Günter Stein
ABSTRACT
NO synthesis is inhibited by the dimethylarginine (DMA) ADMA, which accumulates, similar to SDMA, in the plasma of patients suffering from chronic renal failure (CRF). ADMA and possibly SDMA contribute to hypertension and atherosclerosis in patients with chronic renal disease: ADMA inhibits directly eNOS, whereas SDMA competes with the NO precursor arginine for uptake into the cells. In 26 control persons and 221 patients with kidney diseases of different stage as were CRF, end stage renal disease (ESRD), and patients after renal transplantation (RT), the plasma concentrations of ADMA (c(ADMA)), SDMA (c(SDMA)) and 20 endogenous amino acids (AA) were measured by HPLC and correlated to blood pressure, cardiac events, endothelial dysfunction, and diabetes mellitus. Both ADMA (1.04+/-0.04 vs. 0.66+/-0.04 microM) and SDMA (2.69+/-0.12 vs. 0.49+/-0.03 microM) were significantly (p<0.001) elevated in all patients compared to healthy controls, whereas arginine concentration (51.4+/-2.3 vs. 76.0+/-5.2 microM) was decreased in dependence on the degree of kidney disease. In RT patients, SDMA levels were significantly decreased, but c(ADMA) remained enhanced. A strong correlation was found between SDMA and both serum urea and creatinine in CRF and RT patients. A linear correlation was found between ADMA and cholesterol concentrations in RT patients. Hypertension in CRF was accompanied by a further increase in the concentration of DMAs. There was no relation between DMAs and the occurrence of peripheral arterial occlusive disease or cerebrovascular diseases. In patients with cardiac diseases, c(SDMA) was additionally increased only in the CRF group. In patients with chronic kidney disease, c(ADMA) and c(SDMA) are significantly increased but cardiovascular diseases are evidently not correlated to changes in DMA concentrations in this group of patients.
Sigma-Aldrich
Dimethylamine solution, purum, 33% in absolute ethanol (~5.6 M)
Sigma-Aldrich
3-Hydroxymandelic acid, ≥97.0% (T)
Sigma-Aldrich
Dimethylamine solution, 40 wt. % in H2O
Sigma-Aldrich
Isomaltose, ~98% (TLC)
Sigma-Aldrich
Dimethylamine solution, 2.0 M in methanol
Sigma-Aldrich
1-Methyl-L-histidine, ≥98.0% (TLC)
Sigma-Aldrich
L-2-Aminobutyric acid, ≥99% (titration)
Sigma-Aldrich
L-2-Aminobutyric acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Inosine, ≥99% (HPLC)
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Zinc, powder, <150 μm, 99.995% trace metals basis
Sigma-Aldrich
Zinc, purum, powder
Sigma-Aldrich
Zinc, mossy, ≥99%
Sigma-Aldrich
Trimethylamine N-oxide, 95%
Sigma-Aldrich
Dimethylamine solution, 2.0 M in THF
Sigma-Aldrich
4-Heptanone, 98%
Sigma-Aldrich
4-Heptanone, ≥97%, FG
Sigma-Aldrich
3-Methyl-L-histidine
Sigma-Aldrich
N,N-Dimethylglycine, ≥99%
Sigma-Aldrich
Zinc, granular, 30-100 mesh, 99%
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG
Supelco
4-Heptanone, analytical standard
Sigma-Aldrich
Zinc, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Phenylacetic acid, 99%
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Supelco
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-40, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-200 (10 ampules/kit), 200 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-100 (10 ampules/kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®