Merck
Fluorimetric assay of D-lactate.
Analytical biochemistry (1992-10-01)
A C McLellan, S A Phillips, P J Thornalley
ABSTRACT
A fluorimetric assay for D-lactate in human blood samples was developed using an endpoint enzymatic assay with D-lactate dehydrogenase from Staphylococcus epidermidis. The intrabatch and interbatch coefficients of variance were 8.7% (n = 4) and 16.6% (n = 4), respectively. The limit of detection in blood was 3.73 nmol/ml. The assay suffers minor interference from S-D-lactoylglutathione, which was also present in the blood samples. The concentration of D-lactate in blood was (mean +/- SE, nmol/ml) normal healthy individuals, 11.0 +/- 1.2 (n = 7); and diabetic patients, 20.0 +/- 1.3 (n = 55) (a significant increase in diabetes mellitus; P < 0.01, Mann-Whitney U test).
Product Description
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
1-Butanol, analytical standard
Sigma-Aldrich
Chromium, powder, ≥99% trace metals basis, <45 μm
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Dodecanedioic acid, 99%
Sigma-Aldrich
Chromium, chips, 99.995% trace metals basis
Sigma-Aldrich
Pyruvaldehyde solution, 40 wt. % in H2O
Supelco
Acetone, analytical standard
Sigma-Aldrich
Chromium, powder, 99.5%, −100 mesh
Sigma-Aldrich
1-Butanol, Molecular Biology, ≥99%
Sigma-Aldrich
Chromium, chips, thickness ~2 mm, 99.5%
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
3-Hydroxybutyric acid, 95%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
1-Butanol, 99.9%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
1-Butanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
1-Butanol, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Dimethylamine solution, 2.0 M in methanol
Sigma-Aldrich
L-Carnitine inner salt, synthetic, ≥98%
Sigma-Aldrich
Estriol, ≥97%
Sigma-Aldrich
Estriol, meets USP testing specifications