Merck
Acetaldehyde adducts in the brain of alcoholics.
Archives of toxicology (2003-10-24)
Kazuhiko Nakamura, Kazuhiko Iwahashi, Aizo Furukawa, Kiyoshi Ameno, Hiroshi Kinoshita, Iwao Ijiri, Yoshimoto Sekine, Katsuaki Suzuki, Yasuhide Iwata, Yoshio Minabe, Norio Mori
ABSTRACT
Acetaldehyde binds to some proteins and becomes a Schiff base. It is assumed that after the consumption of ethanol the acetaldehyde binds to the proteins to form adducts, and such acetaldehyde adducts are associated with organ diseases. We investigated the detection of acetaldehyde adducts in the brain region of a human alcoholics. Brain samples collected from an alcoholic autopsied case were used. Determination of acetaldehyde adducts was performed using a fluorescence immmunohistochemical staining method with antibodies against acetaldehyde adducts. We demonstrated acetaldehyde adducts in the frontal cortex and the midbrain of an alcoholics. Our studies showed that an acetaldehyde adduct was produced in the brain of alcoholics.
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
Acetaldehyde solution, 5 M in THF
Sigma-Aldrich
Acetaldehyde solution, 50 wt. % in ethanol
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in isopropanol
Sigma-Aldrich
Acetaldehyde, puriss. p.a., anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Acetaldehyde, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Acetaldehyde, natural, FG
Supelco
Acetaldehyde, PESTANAL®, analytical standard
Sigma-Aldrich
Acetaldehyde, ACS reagent, ≥99.5%