Merck
  • GC-MS determination of flavonoids and phenolic and benzoic acids in human plasma after consumption of cranberry juice.
GC-MS determination of flavonoids and phenolic and benzoic acids in human plasma after consumption of cranberry juice.
Journal of agricultural and food chemistry (2004-01-22)
Kai Zhang, Yuegang Zuo
ABSTRACT
A GC-MS method was developed for the determination of various flavonoids and phenolic and benzoic acids in human plasma. The procedure involved the extraction of flavonoids and phenolic and benzoic acids with ethyl acetate, followed by the derivatization of the phenolic and benzoic compounds with BSTFA (N,O-bis(trimethylsilyl) trifluoroacetamide) + TMCS (trimethylchlorosilane) reagent. The trimethylsilyl derivatives formed were separated and quantitated using GC-MS. Twenty flavonoids and phenolic and benzoic compounds have been well separated in the spiked human plasma without any interference. The average recovery was 79.3%. Several phenolic acids such as o-hydroxybenzoic, p-hydroxyphenylacetic, 2,3-dihydroxybenzoic, 2,4-dihydroxybenzoic, ferulic, sinapic, and benzoic acid were identified and quantified in human plasma after consumption of a cranberry juice. This developed method provides a simple, specific, and sensitive technique for the simultaneous determination of flavonoids and phenolic and benzoic acids in human plasma and is suitable for bioavailability and pharmacokinetic studies.
Product Description
Supelco
Benzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Benzoic acid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Benzoic acid, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
Sigma-Aldrich
Benzoic acid, purified by sublimation, ≥99%
Supelco
Benzoic acid, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
Supelco
Melting point standard 121-123°C, analytical standard
Sigma-Aldrich
Benzoic acid, ≥99.5%, FCC, FG
Salicylic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Salicylic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2,3-Dihydroxybenzoic acid, 99%
Sigma-Aldrich
4-Hydroxyphenylacetic acid, 98%
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Supelco
Salicylic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Supelco
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, (for the calibration of the melting point system), traceable to primary standards (LGC)
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Supelco
Salicylic acid, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®