Merck
  • Obstetric implications of antepartum corticosteroid therapy for HELLP syndrome.
Obstetric implications of antepartum corticosteroid therapy for HELLP syndrome.
Obstetrics and gynecology (2004-11-02)
Carl H Rose, Brad D Thigpen, James A Bofill, Julie Cushman, Warren L May, James N Martin
ABSTRACT
We reviewed the impact of intravenous high-dose corticosteroid administration for preterm hemolysis, elevated liver enzymes, low platelets (HELLP) syndrome on vaginal delivery rate and degree of clinically significant thrombocytopenia. Retrospective analysis of 1991-2000 HELLP syndrome (platelets < 100,000/uL, lactate dehydrogenase > 600 IU/L, aspartate aminotransferase and/or alanine aminotransferase > 70 IU/L) data focusing on labor inductions for gestations of less than 34 weeks and increase in platelet count sufficient to permit regional anesthetic techniques. Antepartum high-dose corticosteroid use increased from 32% (1991-1995) to 67% (1996-2000) for 350 patients studied (n = 199, < 34 weeks; n = 151, > 34 weeks). Corresponding vaginal delivery rates were 32% for gestations of less than 30 weeks, 61% at 30-31 weeks, and 62% at 32-33 weeks. Similarly, 27% of patients with a platelet count of less than 75,000/uL and 52% with a platelet count of less than 100,000/uL who received high-dose corticosteroids during the study interval subsequently achieved a 100,000/uL threshold in time to perform regional anesthesia for delivery. Administration of intravenous high-dose corticosteroids for preterm HELLP syndrome increases probability of successful labor induction and candidacy for regional anesthesia. II-3.
Product Description
Supelco
L-Aspartic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Aspartic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Aspartic acid, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Aspartic acid, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Aspartic acid, BioUltra, ≥99.5% (T)
SAFC
L-Aspartic acid
Sigma-Aldrich
L-Aspartic acid, BioXtra, ≥99% (HPLC)