Merck
  • High glycine levels are associated with prepulse inhibition deficits in chronic schizophrenia patients.
High glycine levels are associated with prepulse inhibition deficits in chronic schizophrenia patients.
Schizophrenia research (2007-02-06)
Uriel Heresco-Levy, Gali Bar, Raz Levin, Marina Ermilov, Richard P Ebstein, Daniel C Javitt
ABSTRACT
Prepulse inhibition (PPI) of the startle reflex, a measure of sensorimotor gating, is decreased in schizophrenia. The validity of a glutamatergic, N-methyl-d-aspartate receptor (NMDAR)-mediated model of PPI disruption is presently equivocal. The NMDAR antagonist ketamine disrupts PPI in rodents, but may increase PPI in healthy volunteers. Glycine (GLY), which acts as an obligatory co-agonist at the NMDAR-GLY site, induces PPI deficits in rats although, consistent with the hypo-NMDAR hypothesis, improves negative and cognitive symptoms in schizophrenia patients. We assessed the hypothesis that GLY serum levels may affect PPI parameters in schizophrenia. Forty-five chronically ill medicated schizophrenia patients and 37 matched healthy comparison subjects were tested for PPI of the eyeblink component of the startle reflex measured by electromyogram recording. Patients' demographic variables, symptom severity scores and GLY, serine and glutamate serum levels were obtained. Patients showed deficient PPI in blocks two and three of the PPI session and differed from controls in terms of change of degree of PPI as a function of the prepulse to eliciting stimulus interval. GLY levels correlated negatively with PPI parameters, such that patients with the highest GLY levels showed decreased PPI (rs=-0.4, p=0.03). These preliminary findings indirectly support previous observations on ketamine effects upon PPI in humans and suggest a dissociation of symptomatology and PPI changes as function of NMDAR modulation in schizophrenia.
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
4-Hydroxyphenylacetic acid, 98%
Supelco
L-Threonine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Tyrosine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
L-Tyrosine, FG
Supelco
L-Aspartic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Alanine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Alanine, ≥99%
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
Homovanillic acid, Fluorimetric reagent
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine solution, 200 mM, Hybri-Max, sterile-filtered, suitable for hybridoma
Sigma-Aldrich
L-Alanine, ≥98% (TLC)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
L-Aspartic acid, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Aspartic acid, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Aspartic acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
L-Tyrosine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Alanine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
SAFC
L-Aspartic acid
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
Sigma-Aldrich
Glycine, tested according to Ph. Eur.