Merck
  • Comparison of oral insulin spray and subcutaneous regular insulin at mealtime in type 1 diabetes.
Comparison of oral insulin spray and subcutaneous regular insulin at mealtime in type 1 diabetes.
Diabetes technology & therapeutics (2007-08-21)
Jaime Guevara-Aguirre, Marco Guevara-Aguirre, Jeannette Saavedra, Gerald Bernstein, Arlan L Rosenbloom
ABSTRACT
The aim of this study was to compare the glucose pharmacodynamics after oral spray insulin (Oral-lyn , Generex Biotechnology, Toronto, ON, Canada) and subcutaneous (sc) injection of regular insulin in 10 subjects with type 1 diabetes mellitus (T1DM). Basal therapy was twice-daily insulin glargine. Preprandial (30 min) regular insulin was given for 3 days, followed by 9 days of Oral-lyn, eight to 12 puffs immediately pre- and postprandially. Adjustments for glycemia were made using standard snacks and additional regular insulin or Oral-lyn. Peripheral glucose measurements were self-monitored in duplicate. Serum concentrations of fructosamine and hemoglobin A1c (HbA1c) were determined at the start and the end of the study period. Average glucose concentrations (in mmol/L) for the 3-day regular insulin and 9-day Oral-lyn periods, respectively, were: pre-breakfast (B), 5.06 and 3.89; 1-h post-B, 8.39 and 7.67; post-B, 6.00 and 6.33; pre-lunch (L), 5.50 and 4.72; 1-h post-L, 7.83 and 7.89; 2-h post-L, 5.89 and 6.33; pre-dinner (D), 5.61 and 5.17; 1-h post-D, 7.22 and 7.83; and 2-h post-D, 6.11 and 6.67. Areas under the curve for both treatments were not significantly different (P = 0.6875). Fructosamine (mean +/- SD, 338.7 +/- 77.4 micromol/L and 321.7 +/- 63.4 micromol/L), and HbA1c (mean +/- SD, 7.5 +/- 1.5% and 7.2 +/- 1.2%) did not change significantly. Regular insulin and Oral-lyn had similar glucodynamic effects in subjects with T1DM receiving twice-daily insulin analogue as baseline therapy. Intensive monitoring and timely corrections with additional snacks, additional sc regular insulin, or Oral-lyn puffs resulted in appropriate glycemic control as assessed by individual daily glycemic responses and, especially, normal preprandial glycemia. Protein glycation decreased, but not significantly.
Sigma-Aldrich
Chromium, chips, thickness ~2 mm, 99.5%
Sigma-Aldrich
Pyruvaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
Chromium, powder, ≥99% trace metals basis, <45 μm
Sigma-Aldrich
Chromium, chips, 99.995% trace metals basis
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Dodecanedioic acid, 99%
Sigma-Aldrich
3-Hydroxybutyric acid, 95%
Supelco
Acetone, analytical standard
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
1-Butanol, suitable for HPLC, ≥99.7%
Sigma-Aldrich
1-Butanol, 99.9%
Sigma-Aldrich
1-Butanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
1-Butanol, Molecular Biology, ≥99%
Supelco
1-Butanol, analytical standard
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Chromium, powder, 99.5%, −100 mesh
Sigma-Aldrich
Dimethylamine solution, 2.0 M in methanol
Supelco
Estriol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Levocarnitine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethylamine solution, purum, 33% in absolute ethanol (~5.6 M)
Sigma-Aldrich
Dimethylamine solution, 2.0 M in THF